Order Entry
Finland
ContactUsLinkComponent
 
 
2-(Bromomethyl)-1-fluoro-3-nitrobenzene 98%
  ACRO449150050
 :  
undefined
 
2-(Bromomethyl)-1-fluoro-3-nitrobenzene 98%
  ACRO449150050
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    Yellow to light brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    50 to 54 °C
  • GC:
    ≥97.5 %

 

 

 :
 :234.02 g/mol
 :50…54 °C
 :Ambient
 :3261
 :8,II
 :GHS05
 :1958-93-6
 :C₇H₅BrFNO₂
 :MFCD00042918