Order Entry
Finland
ContactUsLinkComponent
Calcium acetate xhydrate 99% for analysis
Calcium acetate xhydrate 99% for analysis
Catalog # ACRO447860010
CAS Number:  
undefined
Calcium acetate xhydrate 99% for analysis
Catalog # ACRO447860010
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Appearance:
    White Powder or granules
  • Infrared spectrum:
    Conforms
  • Titration Complexometric:
    ≥98.5 % (on dried substance)
  • Water:
    ≤10 %
  • pH:
    6.0 to 9.0 (10 % in water)
  • Chloride (Cl):
    ≤0.05 %
  • Sulfate (SO4):
    ≤0.1 %
  • Arsenic (As):
    ≤3 ppm
  • Iron (Fe):
    ≤10 ppm
  • Lead (Pb):
    ≤2 ppm
  • Heavy metals (as Pb):
    ≤10 ppm
  • Insoluble matter:
    ≤0.1 %

Specification Test Results

About this item

 :
 :176.18 g/mol
Density:1,5 g/cm³ (20 °C)
Storage Temperature:Ambient
CAS Number:114460-21-8
 :5743-26-0
Formula:(H₃CCOO)₂Ca·nH₂O
MDL Number:MFCD00150019