


N-Methylisopropylamine 98%
:4747-21-1
:
:C₄H₁₁N
:MFCD00042859
:73.14 g/mol
:50…53 °C (760 mmHg)
:0.702 g/cm³
:–32 °C
:2734
:8,I
:GHS02
:GHS05
:GHS06
:Ambient
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3830 to 1.3850 (20°C, 589 nm)
- Titration with HClO4:≥97.5 %