Order Entry
Finland
ContactUsLinkComponent
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS) 98%
Irritant
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS) 98%
Certificates
undefined
Irritant
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS) 98%
Specification Test Results
  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤2 % (K.F.)
  • HPLC:
    ≥97.5 %
  • NMR (1H-NMR):
    Consistent with structure