
3,5-Dichlorothiophenol 97%
:17231-94-6
:
:C₆H₄Cl₂S
:MFCD00041423
:179.07 g/mol
:125…130 °C (12 mmHg)
:62…65 °C
:Ambient
:3335
:9,III
:GHS07
- Appearance:White to yellow Crystalline powder or solid
- Infrared spectrum:Conforms
- Melting point:62 to 65 °C
- GC:≥96.0 %