

2-Amino-3-hydroxybenzoic acid 97%
ACRO434870010
:
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Appearance:Off-white to orange Powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
- GC:≥96.0 %
:
:153.14 g/mol
:357 °C (1013 hPa)
:224…231 °C
:GHS07
:GHS08
:Ambient
:548-93-6
:C₇H₇NO₃
:MFCD00007700