Order Entry
Finland
ContactUsLinkComponent
trans-4-(Trifluoromethyl)cyclohexanecarboxylic acid 98%
Irritant
trans-4-(Trifluoromethyl)cyclohexanecarboxylic acid 98%
Catalog # ACRO432600050
CAS Number:  
undefined
Irritant
trans-4-(Trifluoromethyl)cyclohexanecarboxylic acid 98%
Catalog # ACRO432600050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    152 to 157 °C
  • HPLC:
    ≥97.5 %

Specification Test Results

About this item

 :
 :196.17 g/mol
Melting Pt:153…156 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:133261-33-3
Formula:C₈H₁₁F₃O₂
MDL Number:MFCD09037363