Order Entry
Finland
ContactUsLinkComponent
1-Pentene 97%, AcroSeal®
1-Pentene 97%, AcroSeal®
Catalog # 430731000.
CAS Number:  
undefined
1-Pentene 97%, AcroSeal®
Catalog # 430731000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3704 to 1.3724 (20°C, 589 nm)
  • GC:
    ≥96.0 %

Specification Test Results

About this item

 :
 :70.13 g/mol
Boiling Pt:30 °C (760 mmHg)
Melting Pt:–165 °C
Density:0.64 g/cm³
Flash Pt:–28 °C
UN:1108
ADR:3,I
 :GHS08
 :GHS02
Storage Temperature:Refrigerator
CAS Number:109-67-1
Formula:C₅H₁₀
MDL Number:MFCD00003567