
4-Bromothiazole 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:34259-99-9
:
Formula:C₃H₂BrNS
MDL Number:MFCD06657592
:164.03 g/mol
Flash Pt:102 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless to light brown Liquid
- Infrared spectrum:Conforms
- GC:≥96.0 %