Order Entry
Finland
ContactUsLinkComponent
 
Dicyclohexylcarbodiimide 1 M in dichloromethane, AcroSeal®
Catalog # 429021000.
CAS Number:  
undefined
Dicyclohexylcarbodiimide 1 M in dichloromethane, AcroSeal®
Catalog # 429021000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Solution
  • Infrared spectrum:
    Conforms
  • Titration with HClO4:
    0.95 to 1.10 M
  • GC:
    ≥92 %

Specification Test Results

About this item

 :
Density:1.24 g/cm³
Flash Pt:87 °C
ADR:6.1,III
 :206.33 g/mol
Storage Temperature:Ambient
UN:1593
 :GHS05
 :GHS07
 :GHS08
 :GHS09
CAS Number:538-75-0
Formula:C₁₃H₂₂N₂
MDL Number:MFCD00011659