Order Entry
Finland
ContactUsLinkComponent
Propagyl bromide 80% (w/w) in toluene stabilised, AcroSeal®
Toxic
Health
Corrosive
Flammable
Propagyl bromide 80% (w/w) in toluene stabilised, AcroSeal®
Certificates
undefined
Toxic
Health
Corrosive
Flammable
Propagyl bromide 80% (w/w) in toluene stabilised, AcroSeal®

Specification Test Results

  • Appearance:
    Colorless to yellow-orange Solution
  • Refractive index:
    1.4910 to 1.4970 (20°C, 589 nm)
  • Titration Argentometric:
    75 to 85 % (Product in toluene)
  • Density:
    (20°C) 1.33 to 1.38 g/ml
  • GC:
    ≥96.0 % (Excluding toluene)
  • Stabilizer:
    0.3 % MgO

Specification Test Results