Order Entry
Finland
ContactUsLinkComponent
tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate 98%
Irritant
tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate 98%
Certificates
undefined
Irritant
tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate 98%

Specification Test Results

  • Appearance:
    White to beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    86 to 90 °C
  • HPLC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together