Order Entry
Finland
ContactUsLinkComponent
3-Guanidinopropionic acid 97%
3-Guanidinopropionic acid 97%
Certificates
undefined
3-Guanidinopropionic acid 97%

Specification Test Results

  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    208 to 212 °C
  • NMR (1H-NMR):
    Consistent with structure
  • Titration with HClO4:
    ≥96.0 %

Specification Test Results

Related Information