
4-Bromo-m-anisidine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:19056-40-7
:
Formula:C₇H₈BrNO
MDL Number:MFCD05664063
:202.05 g/mol
Melting Pt:96…100 °C
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:White to brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:95 to 100 °C
- GC:≥96.0 %