Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dibenzyl diisopropylphosphoramidite
Dibenzyl diisopropylphosphoramidite 90+%
Dibenzyl diisopropylphosphoramidite 90+%
:
Thermo Fisher Scientific
Dibenzyl diisopropylphosphoramidite 90+%
:
Thermo Fisher Scientific
:
108549-23-1
:
:
C₂₀H₂₈NO₂P
:
MFCD00191988
:
345.43 g/mol
:
1.028 g/cm³
:
69 °C
:
GHS07
:
Refrigerator
Appearance:
Clear to hazy colorless to light yellow Liquid
Infrared spectrum:
Conforms
GC:
≥90.0 %
ACRO39964