Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Amino-m-cresol
2-Amino-m-cresol 96%
2-Amino-m-cresol 96%
:
Thermo Fisher Scientific
2-Amino-m-cresol 96%
:
Thermo Fisher Scientific
:
2835-97-4
:
:
C₇H₉NO
:
MFCD00075082
:
149…152 °C
:
123.15 g/mol
:
GHS07
:
Ambient
Appearance:
White to brown Crystalline powder or flakes
Infrared spectrum:
Conforms
Melting point:
149 to 152 °C
HPLC:
≥95 %
ACRO39841