Order Entry
Finland
ContactUsLinkComponent
Potassium hydroxide ∼50% in water
Potassium hydroxide ∼50% in water
Catalog # ACRO396060025
CAS Number:  
undefined
Potassium hydroxide ∼50% in water
Catalog # ACRO396060025
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 kg
  • Seq:
    4076

Specifications

Specification Test Results

  • Appearance:
    Colorless Viscous solution
  • Titration with HCl:
    49 to 51 %

Specification Test Results

About this item

 :
 :56.11 g/mol
Boiling Pt:1320 °C (1013 hPa)
Melting Pt:360 °C
Density:1.51 g/cm³
UN:1814
ADR:8,II
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553