Order Entry
Finland
ContactUsLinkComponent
3-Methoxypyridine 97%
Irritant
3-Methoxypyridine 97%
Certificates
undefined
Irritant
3-Methoxypyridine 97%

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5190 to 1.5220 (20°C, 589 nm)
  • GC:
    ≥96.0 %

Specification Test Results

Related Information