
2-Bromo-6-methylpyridine 97%
:5315-25-3
:
:C₆H₆BrN
:MFCD00040743
:172.02 g/mol
:102…103 °C (20 mmHg)
:1.512 g/cm³
:97 °C
:GHS07
:Ambient
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5620 to 1.5640 (20°C, 589 nm)
- GC:≥96.0 %