0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
Wood's alloy (Lipowitz's alloy)
Wood's alloy (lipowitz's alloy), rods
Wood's alloy (lipowitz's alloy), rods
:
Thermo Fisher Scientific
Wood's alloy (lipowitz's alloy), rods
:
Thermo Fisher Scientific
:
76093-98-6
:
8049-22-7
:
MFCD00801077
:
70 °C
:
9,7 g/cm³ (20 °C)
:
Ambient
:
2570
:
6.1,II
:
GHS06
:
GHS08
:
GHS09
Appearance:
Silver grey Rods
Composition:
alloy typically:Bi 50%; Pb 25%; Cd 12.5%; Sn 12.5%
ACRO38855