Order Entry
Finland
ContactUsLinkComponent
 
 
N-(2-Pyridyl)bis(trifluoromethanesulphonimide) 97%
 
undefined
 
N-(2-Pyridyl)bis(trifluoromethanesulphonimide) 97%

 

  • Appearance:
    White to yellow Powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    39 to 43 °C
  • HPLC:
    ≥96.0 %

 

 

Frequently Bought Together