Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Hemin chloride
Hemin (from porcine) ≥99%
Hemin (from porcine) ≥99%
Flammable
Health
Hemin (from porcine) ≥99%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Flammable
Health
Hemin (from porcine) ≥99%
Supplier:
Thermo Fisher Scientific
About this item
CAS Number:
16009-13-5
:
Formula:
C₃₄H₃₂ClFeN₄O₄
MDL Number:
MFCD00010726
:
651.95 g/mol
UN:
3400
ADR:
4.2,III
:
GHS02
:
GHS08
Related Information
Documentation
SDS
ACRO34596
Documentation