Podophyllotoxin 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:518-28-5
:4354-76-1
Formula:C₂₂H₂₂O₈
MDL Number:MFCD00075290
:414.41 g/mol
Melting Pt:178…184 °C
Storage Temperature:Freezer
UN:2811
ADR:6.1,II
:GHS06
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- HPLC:≥94 %