ESD safety ankle boots, lace-up, L10, Vintage Mid
Supplier: ELTEN
Certificates
About this item
Casual, vintage environmental friendly manufactured ESD safety ankle boot made from waxy cowhide leather, a padded tongue plus a low profile, dual-density TPU/PU sole (L10).
- Bioactive fabric lining
- Replaceable insole; orthopaedic insoles possible
- Full length inlay sole (ESD Pro), grey
- Metal-free, puncture resistant midsole
- Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Grey
Certifications: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC.
Specifications
- Catalog No:
- 114-8790
- 114-8797
- 114-8789
- 114-8796
- 114-8795
- 114-8794
- 114-8793
- 114-8792
- 114-8791
- 114-8799
- 114-8798
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001