Accessories for ULT freezer TSX400V
Supplier: Thermo Scientific
Certificates
About this item
Record seven days of temperature performance at temperatures to −150 °C using Thermo Scientific™ chart recorders for ultra-low temperature freezers. Recorders use 150 mm chart paper and are available as factory or field installed.
- Seven-day recorder
- Field or factory installed
- Field-installed units must be installed by a qualified professional
Specifications
- Catalog No:
- 471-1171
- 471-1172
- REVCSS34567
- 471-1173
- HERAFCR567TSX
- Cat. No.:
- 471-1171
- 471-1172
- REVCSS34567
- 471-1173
- HERAFCR567TSX