3-Bromo-2-methyl-1-propene 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1458-98-6
:
Formula:C₄H₇Br
MDL Number:MFCD00134155
Boiling Pt:94…95 °C (760 mmHg)
Density:1.339 g/cm³
Flash Pt:7 °C
UN:2924
ADR:3,II
:135.00 g/mol
:GHS02
:GHS05
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4710 to 1.4730 (20°C, 589 nm)
- GC:≥96.0 %
- Stabilizer:Approx 500 ppm hydroquinone