
2-(3-Aminophenyl)acetic acid 97%
:14338-36-4
:
:C₈H₉NO₂
:MFCD00075058
:151.1626 g/mol
:147…150 °C
:GHS07
:Ambient
- Appearance:Off-white to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:147 to 154 °C
- HPLC:≥96.0 %