Order Entry
Finland
ContactUsLinkComponent
Ammonium sulphide 20% in water
Corrosive
Flammable
Environment
Ammonium sulphide 20% in water
Certificates
undefined
Corrosive
Flammable
Environment
Ammonium sulphide 20% in water
Certificates

Specification Test Results

  • Appearance:
    Clear yellow Solution
  • Titration Iodimetric:
    20.0 to 24.0 %
  • Refractive index:
    1.5290 to 1.5320 (20°C, 589 nm)
  • GC:
    ≥96.0 %

Specification Test Results

Related Information