
4-(Chloromethyl)benzoic acid 98%
ACRO392890500
:
- Pack type:Glass bottle
- Pk:50 g
- Seq:3368
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- Titration with NaOH:≥97.5 %
:
:170.60 g/mol
:202 °C
:Ambient
:3261
:8,II
:GHS05
:1642-81-5
:C₈H₇ClO₂
:MFCD00002568