Allyltrimethoxysilane 97+%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Trimethoxyallylsilane
- CAS Number:2551-83-9
- Formula:H₂CCHCH₂Si(OCH₃)₃
- MDL Number:MFCD00053865
- Molecular Weight:162.26 g/mol
- Flash Pt:46 °C
- Storage Temperature:Ambient
- UN:1993
- ADR:3,III
2 Options Available Below
Product Details & Documents
Synonyms: Trimethoxyallylsilane
- CAS Number:2551-83-9
- Formula:H₂CCHCH₂Si(OCH₃)₃
- MDL Number:MFCD00053865
- Molecular Weight:162.26 g/mol
- Flash Pt:46 °C
- Storage Temperature:Ambient
- UN:1993
- ADR:3,III
Specifications for Product Family
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4040 to 1.4060 (20°C, 589 nm)
- GC:≥97.0 %



