(1S,4S)-2-Boc-2,5-diazabicyclo[2.2.1]heptane 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:113451-59-5
:
Formula:C₁₀H₁₈N₂O₂
MDL Number:MFCD01569250
:198.26 g/mol
Melting Pt:74…76 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light beige Powder
- Infrared spectrum:Conforms
- Melting point:73 to 77 °C
- GC:≥94 %
- Specific optical rotation:-41° to -47° (20 °C, 589 nm) (C=1, chloroform)