
N-Methoxy-N-methylcarbamoyl chloride 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:30289-28-2
:
Formula:C₃H₆ClNO₂
MDL Number:MFCD06658962
:123.54 g/mol
Storage Temperature:Ambient
UN:3265
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4420 to 1.4440 (20°C, 589 nm)
- GC:≥94 %