Order Entry
Finland
ContactUsLinkComponent
N-(tert-Butoxycarbonyl)-trans-4-hydroxy-L-prolinemethyl ester 97%
Irritant
N-(tert-Butoxycarbonyl)-trans-4-hydroxy-L-prolinemethyl ester 97%
Catalog # ACRO376250050
CAS Number:  
undefined
Irritant
N-(tert-Butoxycarbonyl)-trans-4-hydroxy-L-prolinemethyl ester 97%
Catalog # ACRO376250050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to light beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • GC:
    ≥96.0 %
  • Specific optical rotation:
    -63° to -67° (20°C, 589 nm) (c=1, CHCl3)

Specification Test Results

About this item

CAS Number:74844-91-0
 :
Formula:C₁₁H₁₉NO₅
MDL Number:MFCD00076981
Melting Pt:91…96 °C
 :245.28 g/mol
 :GHS07
Storage Temperature:Refrigerator