Tetrahydro-2H-pyran-2-ol 90% cyclized form of 5-Hydroxypentanal
Catalog # ACRO387900250
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- GC:≥88 %
Specification Test Results
About this item
:
:102.133 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:694-54-2
Formula:C₅H₁₀O₂
MDL Number:MFCD02683093