Order Entry
Finland
ContactUsLinkComponent
 
 
Bis(2,2,2-trifluoroethyl) (methoxycarbonylmethyl)phosphonate 90%
  ACRO370610050
 :  
undefined
 
Bis(2,2,2-trifluoroethyl) (methoxycarbonylmethyl)phosphonate 90%
  ACRO370610050
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    Clear orange to dark brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3700 to 1.3740 (20°C, 589 nm)
  • GC:
    ≥88 %

 

 

 :
 :318.11 g/mol
 :1.504 g/cm³
 :>110 °C
 :GHS07
 :Ambient
 :88738-78-7
 :C₇H₉F₆O₅P
 :MFCD00009901