Starch (from potato) for biochemistry, for electrophoresis, hydrolysed
:9005-25-8
:
:(C₆H₁₀O₅)n
:MFCD00082026
:162.14 g/mol
:Ambient
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤20 %
- Gel strength:passes test
- Residue after ignition:≤1 %