Amberlite® XAD4
:9003-69-4
:MFCD00132704
:Ambient
- Appearance:White to yellow Beads
- Particle size:≤5.0 % (< 0.300 mm)
- Harmonic mean size:0.490 to 0.690 mm
- Uniformity coefficient:≤2.0
- Moisture holding capacity:54.0 to 60.0 %
- Ponceau dye test:≤1.0
- Sodium chloride (NaCl):20.0 to 32.0 g/l