Polyethylene glycol monomethyl ether, M.W. ∼350
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9004-74-4
:
Formula:CH₃(OCH₂CH₂)nOH
MDL Number:MFCD00084416
Boiling Pt:>200 °C (760 mmHg)
Density:1.09 g/cm³
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Water:≤0.5 %
- Color scale:≤100 APHA
- pH:4.5 to 7.5 (5% aq. soln., 25°C)
- Hydroxyl value:150 to 172 mg KOH/g