Acrylamide polymer, M.W. M.W. ~ 5 -6.000.000, granules, non-ionic
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9003-05-8
:
Formula:(CH₂:CHCONH₂)n
MDL Number:MFCD00084392
Density:1.3 g/cm³
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light yellow Granules
- Infrared spectrum:Conforms
- Loss on drying:≤13 %
- GC:≥97.5 %