4-Acetyl-4-phenylpiperidine hydrochloride 99%
ACRO134580050
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:Light yellow to beige Powder
- Infrared spectrum:Conforms
- Titration Argentometric:≥98.5 %
- GC:≥98.5 %
:
:239.74 g/mol
:232…234 °C
:Ambient
:10315-03-4
:C₁₃H₁₇NO
:MFCD00039037