Order Entry
Finland
ContactUsLinkComponent
1-(2-Hydroxyethyl)pyrrolidine 95%
1-(2-Hydroxyethyl)pyrrolidine 95%
Certificates
undefined
1-(2-Hydroxyethyl)pyrrolidine 95%

Specification Test Results

  • Appearance:
    Clear light yellow to brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4713 to 1.4743 (20°C, 589 nm)
  • GC:
    ≥94 %

Specification Test Results

Related Information