
Cinnamyl bromide predominantly trans 97%
:4392-24-9
:
:C₉H₉Br
:MFCD00000245
:197.08 g/mol
:103 °C (22 mmHg)
:26…32 °C
:1.332 g/cm³
:87 °C
:Refrigerator
:3261
:8,II
:GHS05
- Appearance:Light yellow to beige to light brown Solidified mass or fragments
- Infrared spectrum:Conforms
- Melting point:26 to 32 °C
- HPLC:≥96.0 %