
9-Acridinecarboxylic acid hydrate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:332927-03-4
Formula:C₁₄H₁₁NO₃
MDL Number:MFCD00149578
:241.24 g/mol
Melting Pt:220 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Bright yellow to yellow-green Crystalline powder
- Infrared spectrum:Conforms
- Water:≤15 % (K.F.)
- Titration with NaOH:≥96.0 % (on dry substance)