Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Morin hydrate
Morin hydrate
Morin hydrate
:
Thermo Fisher Scientific
Morin hydrate
:
Thermo Fisher Scientific
:
654055-01-3
:
:
C₁₅H₁₀O₇
:
MFCD00217054
:
302.24 g/mol
:
300 °C (dec.)
:
3077
:
9,III
:
GHS09
:
GHS08
:
Ambient
Appearance:
Yellow to brown Powder
Infrared spectrum:
Conforms
UV spectrum:
Complies
Water:
≤7 %
Sulfated ash:
≤2 %
ACRO35400