Dodecyl methacrylate 97% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:142-90-5
:
Formula:C₁₆H₃₀O₂
MDL Number:MFCD00008972
:254.41 g/mol
Melting Pt:–23 °C
Density:0.871 g/cm³
Flash Pt:126 °C
Storage Temperature:Ambient
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Appearance:Clear light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4440 to 1.4470 (20°C, 589 nm)
- GC:≥96.0 %
- Stabilizer:300 to 500 ppm MEHQ