
3-Aminopropylphosphonic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13138-33-5
:
Formula:C3H10NO3P
MDL Number:MFCD00008222
:139.0902 g/mol
Melting Pt:290…294 °C (dec.)
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- Titration with NaOH:≥96.0 %