Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Pyrazolidinone hydrochloride
3-Pyrazolidinone hydrochloride 98%
3-Pyrazolidinone hydrochloride 98%
:
Thermo Fisher Scientific
3-Pyrazolidinone hydrochloride 98%
:
Thermo Fisher Scientific
:
1752-88-1
:
MFCD00043787
:
C₃H₇ClN₂O
:
122.552 g/mol
:
GHS07
:
Ambient
Appearance:
White to light yellow Powder
Infrared spectrum:
Conforms
Titration with HClO4:
≥97.5 %
ACRO33549