

2-Methyltetrahydrofuran-3-thiol (cis- and trans- mixture) 97%
Catalog # ACRO332700250
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4730 to 1.4910 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
About this item
:
:118.19 g/mol
Boiling Pt:160…180 °C (760 mmHg)
Density:1.04 g/cm³
Flash Pt:40 °C
UN:1993
ADR:3,III
:GHS07
:GHS02
Storage Temperature:Ambient
CAS Number:57124-87-5
Formula:C₅H₁₀OS
MDL Number:MFCD01632596