Order Entry
Finland
ContactUsLinkComponent
L-Leucinamide hydrochloride 99%
L-Leucinamide hydrochloride 99%
Certificates
undefined
L-Leucinamide hydrochloride 99%
Specification Test Results
  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Titration Argentometric:
    ≥98.5 %
  • Specific optical rotation:
    +9° to +11° (20°C, 589 nm) (c=1, H2O)