Order Entry
Finland
ContactUsLinkComponent
5-Methylfurfurylamine 98%
Irritant
Corrosive
Flammable
5-Methylfurfurylamine 98%
Certificates
undefined
Irritant
Corrosive
Flammable
5-Methylfurfurylamine 98%

Specification Test Results

  • Appearance:
    Clear yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4860 to 1.4880 (20°C, 589 nm)
  • GC:
    ≥97.5 %
  • Titration with HClO4:
    ≥97.5 %

Specification Test Results

Related Information